Learn More
Uvex™ Top Grade 6000 Cowhide Leather Winter Protective Gloves
Catalog Number 15397012
Click to view available options
10
High quality, cowhide leather gloves feature a thick cotton tricot lining for enhanced thermal protection. Gloves offer a good sense of touch, dexterity, and a high level of user comfort.
  • High-quality full cowhide leather winter protection gloves
  • Thick cotton tricot lining increases thermal protection
  • Soft, supple leather ensures dexterity and a good sense of touch
  • For use in dry applications
Colore Gray, Blue
Materiale Cowhide Leather
Size 10
EN388 Standard 3144X
Stile polso guanto Safety
Materiale rivestimento interno Cotton
Serie Top Grade 6000
Lunghezza (sistema metrico) 27 cm