Learn More
Uvex™ Top Grade 6000 Cowhide Leather Winter Protective Gloves
Click to view available options
10
Descrizione
- High-quality full cowhide leather winter protection gloves
- Thick cotton tricot lining increases thermal protection
- Soft, supple leather ensures dexterity and a good sense of touch
- For use in dry applications
Specifica
Colore | Gray, Blue |
Materiale | Cowhide Leather |
Size | 10 |
EN388 Standard | 3144X |
Stile polso guanto | Safety |
Materiale rivestimento interno | Cotton |
Serie | Top Grade 6000 |
Lunghezza (sistema metrico) | 27 cm |