missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trace Amine Receptor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NBP2-24714
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Trace Amine Receptor 1 Polyclonal specifically detects Trace Amine Receptor 1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifica
| Trace Amine Receptor 1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 93% homologous in rhesus monkey, 86% homologous in dog and 50% homologous in mouse and rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin | |
| Q96RJ0 | |
| TAAR1 | |
| A portion of amino acids 200-230 of human TAAR1 was used as the immunogen | |
| 0.1 mg | |
| GPCR | |
| 134864 | |
| Human, Mouse, Rat, Primate | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto