missing translation for 'onlineSavingsMsg'
Learn More

Trace Amine Receptor 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

259.00€ - 452.00€

Specifica

Antigene Trace Amine Receptor 1
Concentrazione 1.0 mg/mL
Diluizione Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin
Applicazioni Western Blot, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
18158251
missing translation for 'viewDocuments'
Novus Biologicals
NBP2-24714SS
0.025 mg
259.00€
25µg
View alternative products
missing translation for 'toSaveUpto' 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
18788963
missing translation for 'viewDocuments'
Novus Biologicals
NBP2-24714
0.1 mg
452.00€
0.10mg
View alternative products
missing translation for 'toSaveUpto' 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Description

Description

Trace Amine Receptor 1 Polyclonal specifically detects Trace Amine Receptor 1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Trace Amine Receptor 1
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human, Mouse, Rat, Primate
Q96RJ0
134864
A portion of amino acids 200-230 of human TAAR1 was used as the immunogen
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
GPCR
PBS with 0.05% Sodium Azide
MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1
TAAR1
IgG
Protein G purified
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 93% homologous in rhesus monkey, 86% homologous in dog and 50% homologous in mouse and rat.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.