missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human CD4 siRNA
Blocks transcription and translation of CD4 from Human.
Marca: enQuireBio™ QX49-2nmol
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000607 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
920 | |
2 nmol | |
Human |
Jurkat Lymphoblast T Cells. | |
CD4 | |
siRNA | |
CD4mut, T-cell surface glycoprotein CD4, T-cell co-receptor CD4 | |
Lyophilized |