Learn More
Uvex Trend 95187 Insole - Width 10
Comfortable climatic insole with antistatic properties for Uvex 2 Trend
Marca: Uvex 9518747
Dettagli aggiuntivi : Peso : 0.10800kg
Descrizione
- Full surface foot shock absorption
- Highly breathable and moisture-absorbent material
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No. 9518.7
- Size: 38 to 52
Specifica
Black | |
Microvelour | |
No | |
Narrow width | |
No |
Uvex 2 Trend | |
47 | |
Unisex | |
Yes |