Learn More
Tricine-SDS Sample Buffer, 2X, reducing, Thermo Scientific™
Click to view available options
Quantità:
25 mL
50 mL
Dimensione della confezione:
25mL
50mL
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Specifica
| Materiale o nome chimico | Sample Loading Buffer |
| Colore | Blue |
| Concentrazione | 2X |
| Forma fisica | Liquid |
| Quantità | 50 mL |