Learn More
TRMT112 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA559026
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: LIQVPKGPVE GYEENEEFLR TMHHLLLEVE VIEGTLQCPE SGRMFPISRG IPNMLLSEEE TES Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
TRMT112 Polyclonal antibody specifically detects TRMT112 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifica
TRMT112 | |
Polyclonal | |
Unconjugated | |
TRMT112 | |
AD-001; HSPC152; HSPC170; hTrm112; Multifunctional methyltransferase subunit TRM112-like protein; TRM112; TRM112-like protein; TRMT112; TRMT11-2; tRNA methyltransferase 112 homolog; tRNA methyltransferase 11-2 homolog (S. cerevisiae) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51504 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UI30 | |
TRMT112 | |
Recombinant protein corresponding to Human TRMT112 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only