missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TST Mouse anti-Human, Mouse, Rat, Clone: 2A6C5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66018-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The product of this gene is a mitochondrial matrix enzyme that is encoded by the nucleus. It may play roles in cyanide detoxification, the formation of iron-sulfur proteins, and the modification of sulfur-containing enzymes. The gene product contains two highly conservative domains (rhodanese homology domains), suggesting these domains have a common evolutionary origin.Specifica
TST | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P24329, P52196, Q16762 | |
TST | |
TST Fusion Protein Ag9235 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
2A6C5 | |
Unconjugated | |
TST | |
RDS, Rhodanese, Thiosulfate sulfurtransferase, TST | |
Mouse | |
Protein A | |
RUO | |
22117, 25274, 7263 | |
-20°C | |
Liquid |