missing translation for 'onlineSavingsMsg'
Learn More

TWF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11732-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899003

  • 121.38€ / 20µL

Spedizione stimata: 29-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.
Specifica

Specifica

TWF1
Polyclonal
Unconjugated
TWF1
A6, Protein A6, Protein tyrosine kinase 9, PTK9, TWF1, Twinfilin 1
Rabbit
Antigen Affinity Chromatography
RUO
19230, 315265, 5756
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q12792, Q5RJR2, Q91YR1
Twf1
TWF1 Fusion Protein Ag2332
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali