missing translation for 'onlineSavingsMsg'
Learn More

TXNDC9 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA556876

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15764781

  • 514.00€ / 100µL
Spedizione stimata: 16-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: TQDYVVGFTD LGNTDDFTTE TLEWRLGSSD ILNYSGNLME PPFQNQKKFG TNFTKLEKKT IRGKKYDSDS DDD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.

Not known. associated with cell differentiation. The protein encoded by this gene is a member of the thioredoxin family. The exact function of this protein is not known but it is associated with cell differentiation.
Specifica

Specifica

TXNDC9
Polyclonal
Unconjugated
TXNDC9
AI098020; APACD; ATP binding protein; ATP binding protein associated with cell differentiation; ATP-binding protein associated with cell differentiation; ES cell-related protein; fb15e07; PHLP3; Protein 1-4; thioredoxin domain containing 9; thioredoxin domain-containing protein 9; TXNDC9; wu:fb15e07; zgc:55522
Rabbit
Antigen affinity chromatography
RUO
10190, 280671, 98258
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O14530, Q8K581, Q9CQ79
TXNDC9
Recombinant protein corresponding to Human TXNDC9
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only