Learn More
Masterflex™ Tygon™ 3 Stop Hydrocarbon Tubing
Tailored for petro-chemicals and resists swelling and cracking. Tygon™ 3 Stop Hydrocarbon Tubing is also UV and Ozone resistant.
Marca: Masterflex™ 95605-40
Dettagli aggiuntivi : Peso : 0.04000kg
Descrizione
- Resists embrittlement
- Compatible with most hydrocarbons
- Resistant to swelling
- Highly flexible, easy to install
- Specifically developed for fuels and lubricants
- Ozone and UV light resistant
Specifica
Green/Green/Green | |
Tygon F4040 | |
12 Pack |
1.85 mm | |
0.85 mm |