missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet Q-CLASSIC-90 Self Closing
Safe and approved storage of flammable hazardous substances in working areas
Marca: asecos™ 34632-041-34637
Dettagli aggiuntivi : Peso : 446.69519kg
Allerta:
For indoor use only
Descrizione
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: door stops in any position, self-closing in the event of fire
- Easy handling with comfort: smooth doors with permanent self-closing function and oil-dampened door closer; open doors with a minimum amout of force
- No unauthorised use: door lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
- Triple certified: cabinet tested in accordance with EN 14470-1, FM Standard 6050 and UL/ULC Standard 1275
Specifica
1647 mm | |
Safety storage cabinet | |
1050 mm | |
Powder-coated textured | |
1193 mm | |
476.50 kg | |
Anthracite Gray, Light Gray | |
615 mm | |
Wing, Self-closing | |
3 | |
2 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | |
Safe and approved storage of flammable hazardous substances in working areas | |
522 mm | |
Steel | |
615 x 1193 x 1953 mm | |
522 x 1050 x 1647 mm | |
Standard | |
1953 mm | |
Q90.195.120.FU.WDC | |
90 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant, FM 6050 approved, UL/ULC 1275 listed |