Learn More
Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-1634-P
Dettagli aggiuntivi : Peso : 0.00100kg
Descrizione
Tyrosinase related Protein 75 (TRP75/gp75) is a pigmentation associated protein expressed by melanoma cells and normal melanocytes. This antigen appears to be expressed during stage III and IV of melanosomes.
Host Species: Mouse
Clone: TA99
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: SK-MEL-23 Melanoma cell line
Molecular Weight: 75kDa
Positive Control: SK-MEL-23, A375 cells; Melanoma, Skin
Cellular Localization: Cytoplasmic Ab-1 is specific for melanomas and does not react with carcinomas and sarcomas.
Recommended for:
- Western Blotting
- Immunohistochemistry (Frozen)
Note:
USA: RUO; Int'l: RUO
Specifica
Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Enzyme | |
Human | |
IgG2a |
Immunohistochemistry (Frozen), Western Blot | |
TA99 | |
Unconjugated | |
SK-MEL-23 Melanoma cell line | |
RUO | |
Primary | |
Concentrated |