Learn More
DuPont™ Tyvek™ Hooded Coverall 800 J, Model TJ 198 Ta
Codice prodotto. p-8349014
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
pezzi 80
Codice prodotto. 15581182
Marca: DuPont™ D15172620
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Achieve an effective barrier against low-concentrated water-based inorganic chemicals and small-sized hazardous particulates, as well as oil repellency using Tyvek™ Impervious Technology. DuPont™ Tyvek™ Hooded Coverall 800 J, Model TJ 198 Ta provides an ideal balance of protection, durability and comfort.
  • Robust yet lightweight (<300g)
  • Self-adhesive zipper flap
  • Self-adhesive chin flap for tight seal of suit to mask
  • Elasticated face, wrists and ankles as well as glued-in waist elastic
  • Thumb loops to prevent sleeves from riding up
  • Available in white, sizes S to XXXL
TRUSTED_SUSTAINABILITY
Specifica
Product Type Hooded Coveralls
Size Small
Materiale Tyvek
Colore White
Monouso Single-use
Stile vita Elastic (Glued-in)
Ankle Style Elastic
Stile polso Elastic, Thumb Loop
Stile collo Con cappuccio
Tipo di chiusura Zipper with Self-adhesive Flap and Chin Flap
Tipo di giuntura Stitched, Taped
Certificazione/Conformità Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B, EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), Antistatic treatment (EN 1149-5) - on inside
Misura petto (sistema metrico) 84 to 92 cm
Da utilizzare con (applicazione) Very humid applications requiring chemical, liquid protection and/or oil repellency
Quantità 25/Pk.
Vedi altri risultati Mostra meno risultati
The user should make the risk assessment to determine the protection required.