Learn More
Uvex™ U-Chem 3100 Chemical Protection Gloves
Catalog Number 17250521
119.70 EUR valido fino al 2024-03-30
Usa il codice promo "21677" per avere il tuo prezzo promozionale.
Click to view available options
Size:
8
9
10
Gloves are constructed of nitrile butadiene rubber (NBR) with a cotton interlock liner, an additional coating for enhances grip capabilities when handling wet or oily objects.
  • Nitrile butadiene rubber (NBR) chemical protection gloves
  • An additional coating enhances grip capabilities when handling wet/oily objects
  • Ideal for a wide variety of applications that require mechanical protection
Applicazioni Laboratory
Coating Material NBR
Colore Black
Size 9
EN388 Standard 4121X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLMO
Da utilizzare con (applicazione) Automotive, Chemical Handling, Metal Work, Mechanics, and Sand Blasting
Sesso Unisex
Stile polso guanto Gauntlet
Serie U-Chem 3100