missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U2AF35 Mouse anti-Human, Clone: 2B7F11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60289-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
U2 auxiliary factor (U2AF), comprising a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. U2 (RNU2) small nuclear RNA auxiliary factor 1 (U2AF1,synonyms: RN, FP793, U2AF35, U2AFBP, RNU2AF1)is the small subunit which plays a critical role in both constitutive and enhancer-dependent RNA splicing by directly mediating interactions between the large subunit and proteins bound to the enhancers. The U2AF1 gene is localized to chromosome 21q22.3, which is the critical region for three diseases, progressive myoclonus epilepsy, autoimmune polyglandular disease type 1, and one form of bipolar affective disorder.Specifica
U2AF35 | |
Monoclonal | |
1.11 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q01081 | |
U2AF1 | |
U2AF35 Fusion Protein Ag0399 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2c |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
2B7F11 | |
Unconjugated | |
U2AF1 | |
FP793, RN, RNU2AF1, U2AF1, U2AF35, U2AFBP | |
Mouse | |
Protein A | |
RUO | |
7307 | |
-20°C | |
Liquid |