missing translation for 'onlineSavingsMsg'
Learn More

U2AF35 Mouse anti-Human, Clone: 2B7F11, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60289-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897355

  • 399.00€ / 150µL
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

U2 auxiliary factor (U2AF), comprising a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. U2 (RNU2) small nuclear RNA auxiliary factor 1 (U2AF1,synonyms: RN, FP793, U2AF35, U2AFBP, RNU2AF1)is the small subunit which plays a critical role in both constitutive and enhancer-dependent RNA splicing by directly mediating interactions between the large subunit and proteins bound to the enhancers. The U2AF1 gene is localized to chromosome 21q22.3, which is the critical region for three diseases, progressive myoclonus epilepsy, autoimmune polyglandular disease type 1, and one form of bipolar affective disorder.
Specifica

Specifica

U2AF35
Monoclonal
1.11 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q01081
U2AF1
U2AF35 Fusion Protein Ag0399
150 μL
Primary
Human
Antibody
IgG2c
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
2B7F11
Unconjugated
U2AF1
FP793, RN, RNU2AF1, U2AF1, U2AF35, U2AFBP
Mouse
Protein A
RUO
7307
-20°C
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali