Learn More
SI Analytics™ Viscometro Ubbelohde
Codice prodotto. 10701761 Sfoglia Tutto SI Analytics Prodotti
Click to view available options
Intervallo viscosità:
0.5 to 3 cSt
0.8 to 5 cSt
1.2 to 10 cSt
10 to 100 cSt
100 to 1000 cSt
1000 to 10,000 cSt
3 to 30 cSt
30 to 300 cSt
5 to 50 cSt
50 to 500 cSt
Costante approssimata:
0.003 K
0.005K
0.01K
0.03K
0.05K
0.1K
0.3K
0.5K
10K
1K
Size:
0.47mm Diameter
0.54mm Diameter
0.63mm Diameter
0.84mm Diameter
0.95mm Diameter
1.13mm Diameter
1.50mm Diameter
1.69mm Diameter
2.01mm Diameter
3.60mm Diameter
Dimensione della confezione:
Pezzo
Codice prodotto. 10701761
Marca: SI Analytics™ 285400172
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Ubbelohde Viscometer includes ring marks to ensure that they can be checked by means of manual measurements.
  • Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
  • The recommended minimum flow through time is 200 seconds
  • Calibrated
  • With constant for automatic measurements
  • Filling quantity of 15 to 20mL
TRUSTED_SUSTAINABILITY
Specifica
Product Type Viscometer
Intervallo viscosità 5 to 50 cSt
Calibrated Yes
Costante approssimata 0.05K
Certificazione/Conformità DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL)
Include Ring Marks
Da utilizzare con (applicazione) For Determination of Absolute and Relative Kinematic Viscosity of Liquids With Newtonian Flow Behavior
Size 0.95mm Diameter
Volume (sistema metrico) 15 to 20 mL