Learn More
SI Analytics™ Viscometro Ubbelohde
Sfoglia Tutto SI Analytics Prodotti
Click to view available options
Intervallo viscosità:
0.5 to 3 cSt
0.8 to 5 cSt
1.2 to 10 cSt
10 to 100 cSt
100 to 1000 cSt
1000 to 10,000 cSt
3 to 30 cSt
30 to 300 cSt
5 to 50 cSt
50 to 500 cSt
Costante approssimata:
0.003 K
0.005K
0.01K
0.03K
0.05K
0.1K
0.3K
0.5K
10K
1K
Size:
0.47mm Diameter
0.54mm Diameter
0.63mm Diameter
0.84mm Diameter
0.95mm Diameter
1.13mm Diameter
1.50mm Diameter
1.69mm Diameter
2.01mm Diameter
3.60mm Diameter
Dimensione della confezione:
Pezzo
Descrizione
- Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
- The recommended minimum flow through time is 200 seconds
- Calibrated
- With constant for automatic measurements
- Filling quantity of 15 to 20mL
Specifica
Specifica
| Product Type | Viscometer |
| Intervallo viscosità | 1000 to 10,000 cSt |
| Calibrated | Yes |
| Costante approssimata | 10K |
| Certificazione/Conformità | DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL) |
| Include | Ring Marks |
| Da utilizzare con (applicazione) | For Determination of Absolute and Relative Kinematic Viscosity of Liquids With Newtonian Flow Behavior |
| Size | 3.60mm Diameter |
| Volume (sistema metrico) | 15 to 20 mL |