missing translation for 'onlineSavingsMsg'
Learn More

UBC9 Mouse anti-Human, Mouse, Rat, Clone: 4A10B11, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60201-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897195

  • 325.38€ / 150µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

In heart and muscle extracts the UBE2I antibodies recognizes a 38 kDa protein band, but this band is not visible in extracts of other rat tissues.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifica

Specifica

UBC9
Monoclonal
1.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P63279, P63280, P63281
Ube2i
UBC9 Fusion Protein Ag7885
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG2a
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry
4A10B11
Unconjugated
UBE2I
C358B7.1, E2,UBE2I, P18, SUMO conjugating enzyme UBC9, SUMO protein ligase, UBC9, UBCE9, UBE2I, Ubiquitin carrier protein 9, Ubiquitin carrier protein I, Ubiquitin protein ligase I
Mouse
Protein A
RUO
22196, 25573, 7329
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali