Learn More
UBTD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13172-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| UBTD2 | |
| Polyclonal | |
| Unconjugated | |
| UBTD2 | |
| DCUBP, DC-UbP, ubiquitin domain containing 2, Ubiquitin like protein SB72, UBTD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 287178, 327900, 92181 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6PGH0, Q8WUN7 | |
| Ubtd2 | |
| UBTD2 Fusion Protein Ag3827 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |