missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12271-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Phosphorylates uridine and cytidine to uridine monophosphate and cytidine monophosphate. Does not phosphorylate deoxyribonucleosides or purine ribonucleosides. Can use ATP or GTP as a phosphate donor. Can also phosphorylate cytidine and uridine nucleoside analogs such as 6-azauridine, 5-fluorouridine, 4- thiouridine, 5-bromouridine, N(4)-acetylcytidine, N(4)- benzoylcytidine, 5-fluorocytidine, 2-thiocytidine, 5- methylcytidine, and N(4)-anisoylcytidine.Specifica
UCK1 | |
Polyclonal | |
Unconjugated | |
UCK1 | |
Cytidine monophosphokinase 1, UCK 1, UCK1, uridine cytidine kinase 1, Uridine monophosphokinase 1, URK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22245, 311864, 83549 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P52623, Q9HA47 | |
UCK1 | |
UCK1 Fusion Protein Ag2912 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |