missing translation for 'onlineSavingsMsg'
Learn More

UFSP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 16999-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16896804

  • 121.38€ / 20µL

Spedizione stimata: 28-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The MW of this protein is 53 kDa, and this antibody specifically recognizes the 53 kDa protein.

Ubiquitin-fold modifier 1 (Ufm1)-specific protease 2 (UfSP2) is a cysteine protease that is responsible for the release of Ufm1 from Ufm1-conjugated cellular proteins, as well as for the generation of mature Ufm1 from its precursor.
Specifica

Specifica

UFSP2
Polyclonal
Unconjugated
UFSP2
C4orf20, UFM1 peptidase 2, Ufm1 specific protease 2, UFSP2
Rabbit
Antigen Affinity Chromatography
RUO
192169, 361151, 55325
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5XIB4, Q99K23, Q9NUQ7
Ufsp2
UFSP2 Fusion Protein Ag10692
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali