Learn More
Erlab™ ULPA U16 Filter
Erlab™ ULPA U16 Filter can be use with a range of Captair Smart filtrating fume hoods and storage cabinets.
Marca: Erlab™ H01074061
Dettagli aggiuntivi : Peso : 2.00000kg
Descrizione
- ULPA U16 filtration efficiency: 99.999995 % according to MPPS method, EN1822 standard
- For use with powders
Specifica
ULPA U16 | |
Filter | |
Powders |