Learn More
Merck Unità di filtrazione centrifughe Ultrafree-CL LG e LH con membrana in PTFE
For use with protein and nucleic acid solutions
281.00€
Specifica
Monouso | Single-use |
---|---|
Materiale (membrana) | PTFE (Hydrophilic) |
Sterilità | Non-sterile |
Materiale alloggiamento | PTFE |
Quantità | 25Pack |
Codice del prodotto | Marca | Colore | Tipo | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Colore | Tipo | Prezzo | Quantità e disponibilità | ||||
10298561
|
Merck
UFC40LG25 |
Yellow | Centrifugal Filters |
281.00€
pezzi 25 |
Spedizione stimata: 04-06-2024
Eseguire il login per visualizzare lo stock disponibile |
||||
Descrizione
Ultrafree™-MC (0.5 mL) and Ultrafree™-CL (2 mL) centrifugal devices are single-use, disposable filters used for removing particulates from aqueous biological solutions. These devices are available in two processing volumes with a range of microporous membranes (from 0.1 to 5.0 μm) for fast filtration and highly reproducible performance. Pre-sterilized units are also available. Use in fixed-angle rotors for 1.5 mL tubes (-MC) and 15 mL tubes (–CL).•Five different pore sizes from 0.1 to 5.0 μm •Pre-sterilized units also available •Fast filtration and highly reproducible performance •Use in fixed-angle rotors for 1.5 mL tubes Applications•Removal of particles and precipitates from aqueous and some solvent-based samples •Ideal for use with protein and nucleic acid solutionsSpecifica
Single-use | |
Non-sterile | |
25Pack | |
0.4 mL | |
16.3 mm |
PTFE (Hydrophilic) | |
PTFE | |
Centrifugal Filters | |
77 cm |