Learn More
Abeba™ UNI6 31763 Shoes
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
Marca: Abeba™ 31763/44
Dettagli aggiuntivi : Peso : 1.14000kg
Descrizione
- CE EN ISO 20345:2011 S1 SRC
- Functional leather
- Dark blue
- Steel toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350122)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Scratch-proof
- **Size 35 with replaceable acc Wave insole (art. 3576)
- Water-repellent
Specifica
CE EN ISO 20345:2011 S1 SRC | |
Leather | |
Slip-resistant | |
31763 |
Blue | |
44 | |
Polyurethane | |
Steel |