Learn More
ABEBA™ Uni6 Black Occupational Trainers O2 ESD
Black Uni6 occupational trainers made of water repellent and breathable fabric, slip resistant PU/TPU outsole
Marca: ABEBA™ 36776/45
Dettagli aggiuntivi : Peso : 0.92000kg
Descrizione
- Comfortable, light occupational shoes
- Comply to EN ISO 20347: 2012, O2, SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant PU/TPU outsole
- Breathable and water repellent knitted fabric
- Breathable and antibacterial lining with silver ions
- Removable insole active comfort
Specifica
Black | |
45 | |
EN ISO 20347: 2012 O2 SRC, ESD according EN ISO 61340 class climatic 1 | |
Trainers | |
O2 SRC ESD |
Knitted Fabric | |
Steel | |
Unisex | |
Water Repellent |