Learn More
Uvex™ Unilite Thermo Plus Cut C Safety Gloves
Winter safety gloves combine protection against thermal risks with level C cut protection. Polymer coating enhances grip and remains flexible even at low temperatures.
Marca:  Uvex™ 6059108
Dettagli aggiuntivi : Peso : 0.12100kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17240461
  • 130.00€ / 10 paia
Spedizione stimata: 09-11-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Winter safety gloves combine thermal protection with level C cut protection
  • Robust polymer coating remains flexible even at low temperatures
  • Acrylic liner provides insulation when in direct contact with cold objects
  • Enhanced grip in dry or slightly damp areas
  • Dual-layer construction
Specifica
Specifica
EN 511: 02X
Polymer
Glass, Polyamide
3X42C
Unisex
Acrylic
Industrial
Black, Green
8
Construction, Forklift Operation, Metal Work, and Refrigeration
Knit
Unilite Thermo Plus Cut C