Learn More
Cytiva Untagged Package
Marca: Cytiva 29-0588-07
Dettagli aggiuntivi : Peso : 0.40000kg
Descrizione
Contents: HiTrap Q HP (1 x 1mL), HiTrap SP HP (1 x 1mL) and HiPrep 16/60 Sephacryl S-200.The Untagged package allows you to combine cation and anion exchange in combination with gel filtration to develop and optimise a purification protocol form most untagged or native proteins.Specifica
Untagged Package | |
HITrap Q 1 x 1 mL, HiTrap SP HP 1 x 1 mL, HiPrep 16/60 Sephacryl S-200 HR | |
Untagged Protein Purification |
2-step protocol, IEX + SEC | |
Ion Exchange Column |