Learn More
Kinesis™ Inlet Solvent Filters
Stainless-steel or multiple materials
Marca: Idex 42711-03
Dettagli aggiuntivi : Peso : 0.06000kg
Descrizione
Cups and end fittings sold separately, see 05-704-54 and 045-707-83.
Specifica
5/Pk. | |
Inlet Solvent Filter | |
0.25 to 28 Coned |
10 μm | |
Stainless Steel | |
40 mL/min. |