Descrizione
Includes:
- Nosepiece 3x bright field, 3x DIC, M27
- FL-LED reflected-light illumination with 4-position reflector turret for P&C modules
- 4-position mount for LED modules
Specifica
LED | |
Upper Stand Part Axio Scope | |
Nosepiece 3x bright field, 3x DIC, M27 - FL-LED reflected-light illumination with 4-position reflector turret for P&C modules and 4-position mount for LED modules |
3X | |
Axio Scope.A1 Microscope | |
3X bright field |