missing translation for 'onlineSavingsMsg'
Learn More

Urea, for Biochemistry, Specially Pure, Fisher Chemical™

Catalog Number 10234690
Click to view available options
Quantità:
1 kg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
57-13-6
CH4N2O
60.056
8022
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
57-13-6
CH4N2O
60.056
8022
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N
Punto di fusione 131°C
Colore White
pH 7.5 to 9.5
Imballaggio Plastic powder jar
Quantità 1 kg
Peso formulazione 60.06g/mol
Pressione del vapore 1.25mmHg 25
Forma fisica Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'