missing translation for 'onlineSavingsMsg'
Learn More

USP1 Mouse anti-Human, Clone: 1C10H6, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66069-1-IG-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897605

  • 121.38€ / 20µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Deubiquitinating enzymes (DUBs) are capable of producing a free ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins in cells. DUBs possess conserved catalytic regions known as the ′cys domain′ and the ′his domain.′ There are 2 distinct families of DUBs: ubiquitin-specific processing proteases (UBPs, or USPs) and ubiquitin C-terminal hydrolases (UCHs).
Specifica

Specifica

USP1
Monoclonal
1.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O94782
USP1
USP1 Fusion Protein Ag5904
20 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
1C10H6
Unconjugated
USP1
Deubiquitinating enzyme 1, hUBP, ubiquitin specific peptidase 1, Ubiquitin thiolesterase 1, UBP, USP1
Mouse
Protein A
RUO
7398
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali