missing translation for 'onlineSavingsMsg'
Learn More

UTP14A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11474-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898573

  • 121.38€ / 20µL

Spedizione stimata: 28-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

UTP14A, a related protein, may also be required for ribosome biogenesis, but not necessarily in a male-specific manner. The gene encoding UTP14C maps to human chromosome 13, which houses over 400 genes, such as BRCA2 and RB1, and comprises nearly 4% of the human genome. As with most chromosomes, polysomy of part or all of chromosome 13 is deleterious to development and decreases the odds of survival. Trisomy 13, also known as Patau syndrome, is deadly and the few who survive past one year suffer from permanent neurologic defects, difficulty eating and vulnerability to serious respiratory infections.
Specifica

Specifica

UTP14A
Polyclonal
Unconjugated
UTP14A
Antigen NY CO 16, KIAA0266, NY CO 16, SDCCAG16, UTP14A
Rabbit
Antigen Affinity Chromatography
RUO
10813, 317579, 72554
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q640M1, Q9BVJ6
Utp14a
UTP14A Fusion Protein Ag2027
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali