missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Safety Glass
17.42 EUR valido fino al 2024-04-30
Usa il codice promo "21673" per avere il tuo prezzo promozionale.
Usa il codice promo "21673" per avere il tuo prezzo promozionale.
Click to view available options
Product Type:
Safety glasses
Descrizione
- Low cost alternative to safety prescription eyewear
- Uvex permanent lens coating technology: anti-fog coating on the inside of the lens, scratch resistant on the outside. For increased durability and safety
- Side arms graduate from hard to soft material, gently grips the side of the head for a secure fit
- Hi resolution lens for improved optical clarity, specifically for wearing two pairs of specs
- 100% UV protection
- Grey sunglare lens – for when sunlight and glare may be a hazard
- Grey sunglare UV 5-2. 5
- Uvex optidur NC
- Black
- EN 166 1F KN CE
Specifica
Certificazione/Conformità | EN 166 1F KN CE |
Product Type | Safety glasses |
Da utilizzare con (applicazione) | Use over most prescription spectacles |
Colore telaio | Black |
N. per confezione | 1 Ea. |
UV Protection | Yes |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'