missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KNF Neuberger™ Aluminium/PTFE Diaphragm Pump
Marca: KNF Neuberger™ 014816/016453
Dettagli aggiuntivi : Peso : 9.02000kg
Specifica
Diaphragms, Valves, Joints, Screws | |
143 mm | |
150 w | |
40°C | |
27 L/min. | |
254 mm | |
Diaphragm Pump |
230 V 50 Hz 150 w | |
Aluminum, PTFE | |
8.2 kg | |
5°C | |
50 Hz | |
265 mm | |
230 V |