Learn More
Labconco™ Vacuum Pump Oil
Manufactured from hydro-treated paraffinic hydrocarbon oil. Labconco™ Vacuum Pump Oil reduces friction, cools and controls contamination when used with general purpose vacuum pumps.
Marca:  Labconco™ 1988000
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 12604576
  • 99.40€ / Pezzo
Spedizione stimata: 13-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Replacement oil for General Purpose Vacuum Pumps 1472100, 7739400, 7739402, 1467700, 7739401 and 7739403
  • Hydro-treated paraffinic hydrocarbon oil
  • One liter