Learn More
Labconco™ Vacuum Pump Oil
Manufactured from hydro-treated paraffinic hydrocarbon oil. Labconco™ Vacuum Pump Oil reduces friction, cools and controls contamination when used with general purpose vacuum pumps.
Marca: Labconco™ 1988000
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Replacement oil for General Purpose Vacuum Pumps 1472100, 7739400, 7739402, 1467700, 7739401 and 7739403
- Hydro-treated paraffinic hydrocarbon oil
- One liter