missing translation for 'onlineSavingsMsg'
Learn More

VAMP3/Cellubrevin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1D10D10, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66488-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898405

  • 399.00€ / 150µL
Spedizione stimata: 16-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Synaptobrevins/VAMPs, syntaxins, and the 25-kDa synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. This gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Because of its high homology to other known VAMPs, its broad tissue distribution, and its subcellular localization, the protein encoded by this gene was shown to be the human equivalent of the rodent cellubrevin. In platelets the protein resides on a compartment that is not mobilized to the plasma membrane on calcium or thrombin stimulation.
Specifica

Specifica

VAMP3/Cellubrevin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P63024, P63025, Q15836
VAMP3
VAMP3/Cellubrevin Fusion Protein Ag1156
150 μL
Primary
Human, Mouse, Porcine, Rat
Antibody
IgG2b
Western Blot
1D10D10
Unconjugated
VAMP3
CEB, Cellubrevin, SYB3, Synaptobrevin 3, VAMP 3, VAMP3
Mouse
Protein A
RUO
100628225, 22319, 29528, 9341
-20°C
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali