Learn More
VAMP8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA552358
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: MEEASEGGGN DRVRNLQSEV EGVKNIMTQN VERILARGEN LEHLRNKTED LEATSEHFKT TSQKVARKFW WKNVKMI Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 92%.
Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.Specifica
VAMP8 | |
Polyclonal | |
Unconjugated | |
VAMP8 | |
AU041171; Edb; endobrevin; hypothetical protein LOC507309; vamp; VAMP8; VAMP-8; vesicle associated membrane protein 8; vesicle-associated membrane protein 8; vesicle-associated membrane protein 8 (endobrevin); wu:fa18d07; zgc:56588 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8673 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BV40 | |
VAMP8 | |
Recombinant protein corresponding to Human VAMP8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only