missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VDAC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55261-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody is specific to VDAC2.
This gene encodes a member of the voltage-dependent anion channel pore-forming family of proteins that are considered the main pathway for metabolite diffusion across the mitochondrial outer membrane. The encoded protein is also thought to be involved in the mitochondrial apoptotic pathway via regulation of BCL2-antagonist/killer 1 protein activity. Pseudogenes have been identified on chromosomes 1, 2, 12 and 21, and alternative splicing results in multiple transcript variants.Specifica
VDAC2 | |
Polyclonal | |
Unconjugated | |
VDAC2 | |
hVDAC2, VDAC 2, VDAC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22334, 7417, 83531 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P45880, P81155, Q60930 | |
VDAC2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |