Learn More
3M™ Versaflo™ Filter TR-3822E
3M™ Versaflo™ Filter TR-3822E for use with 3M™ Versaflo™ TR-300+ Powered Air Respirator.
Marca: 3M™ 7100018861
36.55 EUR valido fino al 2024-12-31
Usa il codice promo "22912" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.20250kg
Descrizione
- P3 particulate filter that provides protection against hydrogen fluoride (HF x 10 TLV) as well as offering nuisance-level acid gas performance
- The filter can be changed quickly and easily by hand
- For use 3M™ TR-300+ Powered Air Respirator
Specifica
HE Filter | |
P3 Particulate, Hydrogen Fluoride, and Nuisance Acid Gas | |
Nuisance-level acid gas relief | |
Belt-mounted |
CE Approved, EN 12941 | |
3M™ Versaflo™ TR-300+ Powered Air Respirator | |
TR-3822E | |
0.352 x 0.2 x 0.13 m |