Learn More
Cytiva (formerly Pall Lab) Versapor™ Acrylic Copolymer 1200 Membrane Disc Filter
Disc Filters useful for prefiltration of particle-laden samples, serum filtration, and dissolution testing.
Marca:  Cytiva (formerly Pall Lab) 66394
Dettagli aggiuntivi : Peso : 0.05100kg
Codice prodotto. 17144561
  • 219.00€ / pezzi 100
Spedizione stimata: 22-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Protects instrumentation against particulate build up
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
Specifica
Versapor (Acrylic Copolymer on a Non-woven Support)
Non-sterile
47 mm
1.2 μm
94 μm