Learn More
Honeywell™ Vertigo Gray PU 3 Gloves
52.50 EUR valido fino al 2024-05-31
Usa il codice promo "21638" per avere il tuo prezzo promozionale.
Usa il codice promo "21638" per avere il tuo prezzo promozionale.
Click to view available options
Size:
8
9
Descrizione
- White Spectra™/lycra™ knitted glove
- Gray polyurethane coating on palm and tips of fingers
- Elastic wrist
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The cool Spectra™ touch gives an added sensation of comfort
- The tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The glove's ventilated back optimises its breathability
- The Spectra®/lycra® thread provides outstanding properties against cuts
- The polyurethane coating at the palm and fingertips provides outstanding abrasion resistance
- Caution! Do not expose this glove to the heat
- Its white color enhances the glove's hygiene and cleanliness
- Its gray coating enables work in a slightly soiled environment
Specifica
Certificazione/Conformità | EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards |
Descrizione | 13 gauge |
Colore | Gray, White |
Materiale | Polyurethane, Lycra |
Size | 9 |
Lunghezza (sistema metrico) | 23.5 cm |