Learn More
VGLUT2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 29209-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The VGLUT2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VGLUT2. This antibody recognizes human, mouse antigen. The VGLUT2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifica
| VGLUT2 | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| DNPI, SLC17A6, VGLUT2 | |
| 20 μL | |
| 57084 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |