Learn More
Macherey-Nagel™ Visocolor™ ECO Test Kit
Allows determination of even low limiting values with accuracy. Macherey-Nagel™ Visocolor™ ECO Test Kits is a group of colorimetric and titrimetric test kits that use precisely dosed single reagents. The results are evaluated visually with high-quality color comparison cards.
Marca: Macherey-Nagel™ 931092
Dettagli aggiuntivi : Peso : 0.30000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Descrizione
Visocolor™ ECO colorimetric and titrimetric test kits achieve high sensitivity and accuracy by using single reagents that can be dosed precisely and by the ability to compensate for the turbidity and color of water samples. The results are evaluated visually with color comparison cards adjusted to the original colors of standard solutions or photometrically with the PF-3 and PF-12Plus photometers for quantitative evaluation.
Easy
- Chemical analysis without further accessories
- No extensive training necessary
- Color-coded reagents with clear dosing instructions
- Easy to understand instruction manual
Safe
- Pictogram test instructions
- Reaction basis according to international standards
- Compensation of turbidity and color
Unique
- High quality test kits
- Business-prized refill packs available
- Ecologically friendly disposal of used reagents
- Delivered in cardboard box with plastic inlay
Specifica
Barium sulfate (turbidity) | |
PF-12 Plus | |
100 Determinations | |
20 to 200mg/L SO42- | |
Colorimetric |
Sulfate determination | |
Reagents, Color comparator, Measuring spoon/glass, Syringe, Instructions in a cardboard box with plastic insert | |
1 Each | |
100 Provisions | |
ECO Sulfate Turbidity Test Kit |
Sicurezza e movimentazione
- VISOCOLOR ECO Sulfate
Avvertenza
- Pericolo
Categoria di pericolo
- Sostanze e miscele corrosive per i metalli Categoria 1
- Tossicità acuta Categoria 3
- Lesione oculare grave/irritazione oculare Categoria 2
Indicazioni di pericolo
- H290-Può essere corrosivo per i metalli.
- H301-Tossico se ingerito.
- H302-Nocivo se ingerito.
- H319-Provoca grave irritazione oculare.
- H332-Nocivo se inalato.
Consigli di prudenza
- P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
- P264-Lavare accuratamente dopo l'uso.
- P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
- P301+P310-IN CASO DI INGESTIONE: contattare immediatamente un CENTRO ANTIVELENI/un medico/
- P301+P312-IN CASO DI INGESTIONE accompagnata da malessere: contattare un CENTRO ANTIVELENI/medico/
- P330-Sciacquare la bocca.
- P390-Assorbire la fuoriuscita per evitare danni materiali.
- P405-Conservare sotto chiave.
Informazioni supplementari
- MIXTURE LIST-Contiene: hydrochloric acid, barium chloride, ammonium chloride