Learn More
VistaLab™ Technologies Ejector Sleeves
For adjustable- and fixed-volume pipetters
Marca: VistaLab™ Technologies 9057-1011
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- ESC: 0.5-20μL, 2-125μL & 5-250μL
- QS & M: 02.-2μL, 1-10μL, 2-20μL, 10-100μL, 20-200μL
- F1 & F2: all models dispensing 20oμL and less
Specifica
Electronic Single Channel Model 0.5 to 20 μL, 2 to125 μL, 5 to 250 μL Model, All Adjustable Models (Except 100 to 1000 μL), All Fixed Volume Models Dispensing 200 μL and | |
5 in. | |
1 in. |
1 Ea. | |
3 in. |