missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitlab™ VITsafe™ Narrow-mouth Safety Wash Bottles
Click to view available options
Capacità (sistema metrico):
250 mL
Descrizione
- Bottles made from PE-LD, spray tube from PP.
- With colored VENT-CAP which supports easy visual identification of the media.
- Chemical formula, CAS No., hazard pictogram, signal word
- Risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
Specifica
Diametro (sistema metrico) | 65 mm |
Altezza (sistema metrico) | 135 mm |
Chemical Label | Methanol |
Capacità (sistema metrico) | 250 mL |
Materiale | LDPE |
N. per confezione | 12/PK |
Dimensioni filettatura (sistema metrico) | GL 25 |
Colore chiusura | Green |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'