Learn More
Emma Safety Footwear Vulcanus XD Safety Shoes
Welder's boot designed specifically for working with metal in extreme temperatures. Over-foot flap provides no entry for sparks or shards.
Marca: Emma Safety Footwear MM54086845
Dettagli aggiuntivi : Peso : 2.08700kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Velcro over-foot flap
- Sanitized
Specifica
Vulcanus XD | |
Leather | |
45 |