Learn More
VWA3A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA559678
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: QGIYLFTGGI PDQDMPTLSA YMAEACGGCD LQLNVCLFYV GEPKMDTTPP ARYASHTDTA AAYKEVTRAA GGRFHWFGDT GIYESDDINS IMSEMEKALN YSQKCA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
VWA3A Polyclonal antibody specifically detects VWA3A in Human samples. It is validated for Immunohistochemistry.Specifica
VWA3A | |
Polyclonal | |
Unconjugated | |
VWA3A | |
von Willebrand factor A domain containing 3A; von Willebrand factor A domain-containing protein 3A; VWA3A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
146177 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NCI4 | |
VWA3A | |
Recombinant protein corresponding to Human VWA3A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only