missing translation for 'onlineSavingsMsg'
Learn More
Learn More
vwf Mouse anti-Human, Clone: 3F9F3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66682-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.Specifica
vwf | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P04275 | |
VWF | |
vwf Fusion Protein Ag25578 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin) | |
3F9F3 | |
Unconjugated | |
VWF | |
F8VWF, von Willebrand antigen II, von Willebrand factor, VWD, VWF | |
Mouse | |
Protein G | |
RUO | |
7450 | |
-20°C | |
Liquid |